missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kif4A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35106-100ul
This item is not returnable.
View return policy
Description
Kif4A Polyclonal antibody specifically detects Kif4A in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| Kif4A | |
| Polyclonal | |
| Western Blot 1:1000 - 1:3000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| chromokinesin-A, chromosome-associated kinesin KIF4A, FLJ12530, FLJ12655, FLJ14204, FLJ20631, HSA271784, KIF4G1, KIF4KIF4-G1, kinesin family member 4A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 870-1080 of human Kif4A (NP_036442.3).,, Sequence:, LIGELVSSKIQVSKLESSLKQSKTSCADMQKMLFEERNHFAEIETELQAELVRMEQQHQEKVLYLLSQLQQSQMAEKQLEESVSEKEQQLLSTLKCQDEELEKMREVCEQNQQLLRENEIIKQKLTLLQVASRQKHLPKDTLLSPDSSFEYVPPKPKPSRVKEKFLEQSMDIEDLKYCSEHSVNEHEDGDGDDDEGDDEEWKPTKLVKVSR | |
| 100 μL | |
| Core ESC Like Genes, Lipid and Metabolism, Mitotic Regulators, Neuroscience, Stem Cell Markers | |
| 24137 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction