missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF3B Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Specifications
| Antigen | KIF3B |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18306793
|
Novus Biologicals
NBP3-05143-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18368642
|
Novus Biologicals
NBP3-05143-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KIF3B Polyclonal specifically detects KIF3B in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| KIF3B | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 9371 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| HH0048, KIAA0359Microtubule plus end-directed kinesin motor 3B, kinesin family member 3B, kinesin-like protein KIF3B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 663-747 of human KIF3B (NP_004789.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title