missing translation for 'onlineSavingsMsg'
Learn More

KIF3A Antibody (SR2233), Novus Biologicals™

Rabbit Monoclonal Antibody

£257.00 - £341.00

Specifications

Antigen KIF3A
Clone SR2233
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 1:50
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18652725
View Documents
Novus Biologicals
NBP3-21780-100ul
100 μg
£341.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18626834
View Documents
Novus Biologicals
NBP3-21780-50ul
50 μg
£257.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

KIF3A Monoclonal antibody specifically detects KIF3A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications

Specifications

KIF3A
Western Blot 1:500-1:2000, Immunoprecipitation 1:50
Monoclonal
Purified
RUO
Human, Mouse, Rat
KIF3, kinesin family member 3A, kinesin family protein 3A, kinesin-like protein KIF3A, Microtubule plus end-directed kinesin motor 3A
A synthesized peptide derived from human KIF3A (Uniprot #: Q9Y496)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR2233
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Cell Biology, Cytoskeleton Markers, Immunology, Neuroscience, Signal Transduction, Stem Cells
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
11127
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.