missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF2C Rabbit anti-Human, Clone: 7V9D2, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | KIF2C |
|---|---|
| Clone | 7V9D2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18315054
|
Novus Biologicals
NBP3-16764-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18355024
|
Novus Biologicals
NBP3-16764-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KIF2C Monoclonal antibody specifically detects KIF2C in Human samples. It is validated for Western BlotSpecifications
| KIF2C | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cellular Markers, DNA Repair | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 11004 | |
| IgG | |
| Affinity purified |
| 7V9D2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| kinesin family member 2C, kinesin-like 6 (mitotic centromere-associated kinesin), Kinesin-like protein 6, kinesin-like protein KIF2C, MCAKKNSL6kinesin-like 6, Mitotic centromere-associated kinesin | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human KIF2C/KIF2C (Q99661). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title