missing translation for 'onlineSavingsMsg'
Learn More

KIF2C Antibody (KIF2C/6529), Novus Biologicals™

Mouse Monoclonal Antibody

£195.00 - £385.00

Specifications

Antigen KIF2C
Clone KIF2C/6529
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18624864
View Documents
Novus Biologicals
NBP3-20514-100ug
100 μg
£385.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18661335
View Documents
Novus Biologicals
NBP3-20514-20ug
20 μg
£195.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

KIF2C Monoclonal antibody specifically detects KIF2C in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

KIF2C
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cancer, Cell Biology, Cellular Markers, DNA Repair
10mM PBS with 0.05% BSA
11004
IgG2b κ
Protein A or G purified
KIF2C/6529
Immunohistochemistry-Paraffin 1-2 μg/mL
Monoclonal
Purified
RUO
Human
kinesin family member 2C, kinesin-like 6 (mitotic centromere-associated kinesin), Kinesin-like protein 6, kinesin-like protein KIF2C, MCAKKNSL6kinesin-like 6, Mitotic centromere-associated kinesin
Recombinant fragment (around aa500-700) of human KIF2C protein (exact sequence is proprietary)
Primary
Store at 4&drg;C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.