missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13711-1-AP-20UL
This item is not returnable.
View return policy
Description
KIF2B is the plus end-directed microtubule-dependent motor required for spindle assembly and chromosome movement. KIF2B has microtubule depolymerization activity.Specifications
| KIF2B | |
| Polyclonal | |
| Unconjugated | |
| KIF2B | |
| KIF2B, kinesin family member 2B, Kinesin like protein KIF2B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 287624, 73470, 84643 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5XI51, Q8C0N1, Q8N4N8 | |
| Kif2b | |
| KIF2B Fusion Protein Ag4648 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction