missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIF2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555186
This item is not returnable.
View return policy
Description
Immunogen sequence: LPESPCLSPL KPLKPHFGDI QEGIYVAIQR SDKRIHLAVV TEINRENYWV TVEWVEKAVK KGKKIDLETI LLLNPALDSA EHP Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.
KIF2B is the plus end-directed microtubule-dependent motor required for spindle assembly and chromosome movement. KIF2B has microtubule depolymerization activity.
Specifications
| KIF2B | |
| Polyclonal | |
| Unconjugated | |
| KIF2B | |
| 1700063D03Rik; EMBL:AAH83841.1, ECO:0000312; KIF2B; kif2b {ECO:0000312; kinesin family member 2B; kinesin protein; kinesin-like protein KIF2B; LOW QUALITY PROTEIN: kinesin-like protein KIF2B; RGD:1359248}; testis tissue sperm-binding protein Li 82P | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84643 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.30 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N4N8 | |
| KIF2B | |
| Recombinant protein corresponding to Human KIF2B. Recombinant protein control fragment (Product #RP-93276). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction