missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF2A Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13105-1-AP-20UL
This item is not returnable.
View return policy
Description
Kif2a gene ontology annotations related to this gene include microtubule-based movement.Specifications
| KIF2A | |
| Polyclonal | |
| Unconjugated | |
| KIF2A | |
| HK2, KIF2, KIF2A, Kinesin 2, kinesin heavy chain member 2A, Kinesin like protein KIF2A, KNS2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16563, 3796, 478071, 84391 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O00139, P28740, Q9WV63 | |
| Kif2a | |
| KIF2A Fusion Protein Ag3748 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse, Canine | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction