missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KIF23 Recombinant Rabbit Monoclonal Antibody (K01_1T93)
GREENER_CHOICE

Product Code. 30829359
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829359 100 μL
30828847 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829359 Supplier Invitrogen™ Supplier No. MA563669
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

KIF23 Recombinant Monoclonal Antibody for Western Blot

The protein encoded by this gene is a member of kinesin-like protein family. This family includes microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. This protein has been shown to cross-bridge antiparallel microtubules and drive microtubule movement in vitro. Alternate splicing of this gene results in two transcript variants encoding two different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF23
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1T93
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene KIF23
Gene Accession No. Q02241
Gene Alias 3110001D19Rik; C87313; cb738; CHO1; KIF23; kif23 protein; kinesin family member 23; kinesin-like 5; kinesin-like 5 (mitotic kinesin-like protein 1); Kinesin-like protein 5; Kinesin-like protein KIF23; Kinesin-like protein KIF23-like protein; knsl5; Mitotic kinesin-like protein 1; mklp1; MKLP-1; wu:fi30e02; wu:fi39f08; zMKLP1
Gene Symbols KIF23
Host Species Rabbit
Immunogen A synthetic peptide of human MKLP1.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9493
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.