missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KIF22 Polyclonal antibody specifically detects KIF22 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | KIF22 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Formulation | PBS, pH 7.2, 40% glycerol |
| Gene Alias | Kid, KIDA-328A3.2, kinesin family member 22, kinesin-like 4, Kinesin-like DNA-binding protein, Kinesin-like protein 4, kinesin-like protein KIF22, KNSL4kinesin-like DNA-binding protein pseudogene, OBP, OBP-1, OBP-2, origin of plasmid DNA replication-binding protein, oriP binding protein |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: LEPEEKAEDCWELQISPELLAHGRQKILDLLNEGSARDLRSLQRIGPKKAQLIVGWRELHGPFSQVEDLERVEGITGKQMESF |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?