missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF22 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35523-100ul
This item is not returnable.
View return policy
Description
KIF22 Polyclonal antibody specifically detects KIF22 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| KIF22 | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Kid, KIDA-328A3.2, kinesin family member 22, kinesin-like 4, Kinesin-like DNA-binding protein, Kinesin-like protein 4, kinesin-like protein KIF22, KNSL4kinesin-like DNA-binding protein pseudogene, OBP, OBP-1, OBP-2, origin of plasmid DNA replication-binding protein, oriP binding protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 456-665 of human KIF22 (NP_015556.1).,, Sequence:, QGAPLLSTPKRERMVLMKTVEEKDLEIERLKTKQKELEAKMLAQKAEEKENHCPTMLRPLSHRTVTGAKPLKKAVVMPLQLIQEQAASPNAEIHILKNKGRKRKLESLDALEPEEKAEDCWELQISPELLAHGRQKILDLLNEGSARDLRSLQRIGPKKAQLIVGWRELHGPFSQVEDLERVEGITGKQMESFLKANILGLAAGQRCGAS | |
| 100 μL | |
| Core ESC Like Genes, Phospho Specific, Stem Cell Markers | |
| 3835 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction