Learn More
KIF20A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15911-1-AP-150UL
 View more versions of this product
Product Code. 16885304
  • £349.00 / 150µL
Description
This gene encodes a glycoprotein expressed on the surface of NK cells, platelets, monocytes and a subset of T cells. It is a member of the Ig-superfamily containing 2 Ig-like domains of the V-set. The protein mediates cellular adhesion of platelets and megakaryocytic cells to vascular endothelial cells. The protein also plays a role in megakaryocytic cell maturation.
Specifications
Specifications
KIF20A
Polyclonal
Unconjugated
KIF20A
GG10_2, KIF20A, kinesin family member 20A, Kinesin like protein KIF20A, MKLP2, RAB6KIFL, Rabkinesin 6
Rabbit
Antigen Affinity Chromatography
RUO
10112, 19348, 361308
-20°C
Liquid
Immunoprecipitation, Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95235, P97329
Kif20a
KIF20A Fusion Protein Ag8700
150 μL
Primary
Human, Mouse, Rat