missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF20A, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00010112-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MSQGILSPPAGLLSDDDVVVSPMFESTAADLGSVVRKNLLSDCSVVSTSLEDKQQVPSEDSMEKVKVYLRVRPLLPSELERQEDQGCVRIENVETLVLQAPKDSFALKSNERGIGQATHRFTFSQIFGPEVGQASFFNLTVKEMVKDVLKGQNWLIYTYGVTNSGKTHTIQGTIKDGGILPRSLALIFNSLQGQLHPTPDLKPLLSNEVIWLDSKQIRQEEMKKLSLLNGGLQEEELSTSLKRSVYIESRIGTSTSFDSGIAGLSSISQCTSSSQLDETSHRWAQPDTAPLPVPANIRFSIWISFFEIYNELLYDLLEPPSQQRKRQTLRLCEDQNGNPYVKDLNWIHVQDAEEAWKLLKVGRKNQSFASTHLNQNSSRSHSIFSIRILHLQGEGDIVPKISELSLCDLAGSERCKDQKSGERLKEAGNINTSLHTLGRCIAALRQNQQNRSKQNLVPFRDSKLTRVFQGFFTGRGRSCMIVNVNPCASTYDETLHVAKFSAIASQLVHAPPMQLGFPSLHSFIKEHSLQVSPSLEKGAKADTGLDDDIENEADISMYGKEELLQVVEAMKTLLLKERQEKLQLEMHLRDEICNEMVEQMQQREQWCSEHLDTQKELLEEMYEEKLNILKESLTSFYQEEIQERDEKIEELEALLQEARQQSVAHQQSGSELALRRSQRLAASASTQQLQEVKAKLQQCKAELNSTTEELHKYQKMLEPPPSAKPFTIDVDKKLEEGQKNIRLLRTELQKLGESLQSAERACCHSTGAGKLRQALTTCDDILIKQDQTLAELQNNMVLVKLDLRKKAACIAEQYHTVLKLQGQVSAKKRLGTNQENQQPNQQPPGKKPFLRNLLPRTPTCQSSTDCSPYARILRSRRSPLLKSGPFGKKYSpecifications
| KIF20A | |
| Polyclonal | |
| No additive | |
| NM_005733 | |
| KIF20A | |
| KIF20A (NP_005724.1, 1 a.a. ∼ 890 a.a) full-length human protein. | |
| RUO | |
| 10112 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| kinesin family member 20A | |
| FLJ21151/MKLP2/RAB6KIFL | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction