missing translation for 'onlineSavingsMsg'
Learn More

KIF20A Antibody, Novus Biologicals™

Product Code. 18339139 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339139 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18339139 Supplier Novus Biologicals Supplier No. H00010112B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

KIF20A Polyclonal antibody specifically detects KIF20A in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF20A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_005724
Gene Alias FLJ21151, GG10_2, kinesin family member 20A, kinesin-like protein KIF20A, mitotic kinesin-like protein 2, MKLP2, Rab6-interacting kinesin-like protein, RAB6KIFLkinesin-like (rabkinesin6), rabkinesin-6
Host Species Mouse
Immunogen KIF20A (NP_005724, 1 a.a. - 890 a.a.) full-length human protein. MSQGILSPPAGLLSDDDVVVSPMFESTAADLGSVVRKNLLSDCSVVSTSLEDKQQVPSEDSMEKVKVYLRVRPLLPSELERQEDQGCVRIENVETLVLQAPKDSFALKSNERGIGQATHRFTFSQIFGPEVGQASFFNLTVKEMVKDVLKGQNWLIYTYGVTNSGKTHTIQGTIKDGGILPRSLALIFNSLQGQLHPTPDLKPLLSNEVIWLDSKQIRQEEMKKLSLLNGGLQEEELSTSLKRSVYIESRIGTSTSFDSGIAGLSSISQCTSSSQLDETSHRWAQPDTAPLPVPANIRFSIWISFFEIYNELLYDLLEPPSQQRKRQTLRLCEDQNGNPYVKDLNWIHVQDAEEAWKLLKVGRKNQSFASTHLNQNSSRSHSIFSIRILHLQGEGDIVPKISELSLCDLAGSERCKDQKSGERLKEAGNINTSLHTLGRCIAALRQNQQNRSKQNLVPFRDSKLTRVFQGFFTGRGRSCMIVNVNPCASTYDETLHVAKFSAIASQLVHAPPMQLGFPSLHSFIKEHSLQVSPSLEKGAKADTGLDDDIENEADISMYGKEELLQVVEAMKTLLLKERQEKLQLEMHLRDEICNEMVEQMQQREQWCSEHLDTQKELLEEMYEEKLNILKESLTSFYQEEIQERDEKIEELEALLQEARQQSVAHQQSGSELALRRSQRLAASASTQQLQEVKAKLQQCKAELNSTTEELHKYQKMLEPPPSAKPFTIDVDKKLEEGQKNIRLLRTELQKLGESLQSAERACCHSTGAGKLRQALTTCDDILIKQDQTLAELQNNMVLVKLDLRKKAACIAEQYHTVLKLQGQVSAKKRLGTNQENQQPNQQPPGKKPFLRNLLPRTPTCQSSTDCSPYARILRSRRSPLLKSGPFGKKY
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10112
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.