missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIF20A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144265
This item is not returnable.
View return policy
Description
KIF20A Polyclonal Antibody for IHC (P), IP, ELISA
Mitotic kinesin required for chromosome passenger complex (CPC)-mediated cytokinesis. Following phosphorylation by PLK1, involved in recruitment of PLK1 to the central spindle. Interacts with guanosine triphosphate (GTP)-bound forms of RAB6A and RAB6B. May act as a motor required for the retrograde RAB6 regulated transport of Golgi membranes and associated vesicles along microtubules. Has a microtubule plus end-directed motility. [UniProt].
Specifications
| KIF20A | |
| Polyclonal | |
| Unconjugated | |
| KIF20A | |
| AA415432; GG10_2; KIF20A; kinesin family member 20A; kinesin-like protein 174; Kinesin-like protein KIF20A; mitotic kinesin-like protein 2; MKlp2; RAB6 interacting, kinesin-like (rabkinesin6); rab6-interacting kinesin-like protein; RAB6KIFL; Rabkinesin-6 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 10112 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 3.55 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| O95235 | |
| KIF20A | |
| Recombinant Human Kinesin-like protein KIF20A protein (161-294AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction