missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF19 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19551-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody recognizes isoform1 of KIF19 only.
Plus end-directed microtubule-dependent motor protein that regulates the length of motile cilia by mediating depolymerization of microtubules at ciliary tips.Specifications
| KIF19 | |
| Polyclonal | |
| Unconjugated | |
| KIF19 | |
| KIF19, KIF19A, kinesin family member 19, Kinesin like protein KIF19 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 124602, 286942 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.57 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q2TAC6, Q99PT9 | |
| KIF19 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction