missing translation for 'onlineSavingsMsg'
Få mere at vide

KIF18A Rabbit anti-Human, Polyclonal, Novus Biologicals™

Artikelnummer. 18344152 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
20 μL
100 μL
Pakningsstørrelse:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
18344152 20 μL 20µL
18379751 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18344152 Leverandør Novus Biologicals Leverandørnr. NBP30512020UL

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

KIF18A Polyclonal specifically detects KIF18A in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen KIF18A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias DKFZP434G2226, kinesin family member 18A, kinesin-like protein KIF18A, Marrow stromal KIF18A, MS-KIF18A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 599-898 of human KIF18A (NP_112494.3).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81930
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.