missing translation for 'onlineSavingsMsg'
Learn More

KIF18A Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18379751 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379751 100 μL 100µL
18344152 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18379751 Supplier Novus Biologicals Supplier No. NBP305120100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KIF18A Polyclonal specifically detects KIF18A in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF18A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias DKFZP434G2226, kinesin family member 18A, kinesin-like protein KIF18A, Marrow stromal KIF18A, MS-KIF18A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 599-898 of human KIF18A (NP_112494.3).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81930
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.