missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF16B Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Specifications
| Antigen | KIF16B |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18393937
|
Novus Biologicals
NBP3-04523-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18323107
|
Novus Biologicals
NBP3-04523-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KIF16B Polyclonal specifically detects KIF16B in Human samples. It is validated for Western Blot.Specifications
| KIF16B | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| C20orf23, chromosome 20 open reading frame 23, dJ971B4.1, FLJ20135, KIAA1590, kinesin family member 16B, kinesin-like protein KIF16B, KISC20ORF, SNX23kinesin motor protein, sorting nexin 23, Sorting nexin-23 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human KIF16B (NP_078980.3). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 55614 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title