missing translation for 'onlineSavingsMsg'
Learn More

KIF16B Antibody, Novus Biologicals™

Product Code. 18362109 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18362109 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18362109 Supplier Novus Biologicals Supplier No. H00055614B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

KIF16B Polyclonal antibody specifically detects KIF16B in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF16B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. NP_078980
Gene Alias C20orf23, chromosome 20 open reading frame 23, dJ971B4.1, FLJ20135, KIAA1590, kinesin family member 16B, kinesin-like protein KIF16B, KISC20ORF, SNX23kinesin motor protein, sorting nexin 23, Sorting nexin-23
Host Species Mouse
Immunogen C20orf23 (NP_078980, 1 a.a. - 1317 a.a.) full-length human protein. MASVKVAVRVRPMNRREKDLEAKFIIQMEKSKTTITNLKIPEGGTGDSGRERTKTFTYDFSFYSADTKSPDYVSQEMVFKTLGTDVVKSAFEGYNACVFAYGQTGSGKSYTMMGNSGDSGLIPRICEGLFSRINETTRWDEASFRTEVSYLEIYNERVRDLLRRKSSKTFNLRVREHPKEGPYVEDLSKHLVQNYGDVEELMDAGNINRTTAATGMNDVSSRSHAIFTIKFTQAKFDSEMPCETVSKIHLVDLAGSERADATGATGVRLKEGGNINKSLVTLGNVISALADLSQDAANTLAKKKQVFVPYRDSVLTWLLKDSLGGNSKTIMIATISPADVNYGETLSTLRYANRAKNIINKPTINEDANVKLIRELRAEIARLKTLLAQGNQIALLDSPTALSMEEKLQQNEARVQELTKEWTNKWNETQNILKEQTLALRKEGIGVVLDSELPHLIGIDDDLLSTGIILYHLKEGQTYVGRDDASTEQDIVLHGLDLESEHCIFENIGGTVTLIPLSGSQCSVNGVQIVEATHLNQGAVILLGRTNMFRFNHPKEAAKLREKRKSGLLSSFSLSMTDLSKSRENLSAVMLYNPGLEFERQQREELEKLESKRKLIEEMEEKQKSDKAELERMQQEVETQRKETEIVQLQIRKQEESLKRRSFHIENKLKDLLAEKEKFEEERLREQQEIELQKKRQEEETFLRVQEELQRLKELNNNEKAEKFQIFQELDQLQKEKDEQYAKLELEKKRLEEQEKEQVMLVAHLEEQLREKQEMIQLLRRGEVQWVEEEKRDLEGIRESLLRVKEARAGGDEDGEELEKAQLRFFEFKRRQLVKLVNLEKDLVQQKDILKKEVQEEQEILECLKCEHDKESRLLEKHDESVTDVTEVPQDFEKIKPVEYRLQYKERQLQYLLQNHLPTLLEEKQRAFEILDRGPLSLDNTLYQVEKEMEEKEEQLAQYQANANQLQKLQATFEFTANIARQEEKVRKKEKEILESREKQQREALERALARLERRHSALQRHSTLGMEIEEQRQKLASLNSGSREQSGLQASLEAEQEALEKDQERLEYEIQQLKQKIYEVDGVQKDHHGTLEGKVASSSLPVSAEKSHLVPLMDARINAYIEEEVQRRLQDLHRVISEGCSTSADTMKDNEKLHNGTIQRKLKYERMVSRSLGANPDDLKDPIKISIPRYVLCGQGKDAHFEFEVKITVLDETWTVFRRYSRFREMHKTLKLKYAELAALEFPPKKLFGNKDERVIAERRSHLEKYLRDFFSVMLQSATSPLHINKVGLTLSKHTICEFSPFFKKGVFDYSSHGTG
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55614
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.