Learn More
KIF15 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30774-1-AP-20UL
This item is not returnable.
View return policy
Description
The KIF15 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KIF15. This antibody recognizes human antigen. The KIF15 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| KIF15 | |
| 700 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| FLJ25667, HKLP2, KIF15, kinesin family member 15, Kinesin like protein 2, Kinesin like protein 7, Kinesin like protein KIF15, KLP2, KNSL7, NY BR 62 | |
| 20 μL | |
| 56992 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |