missing translation for 'onlineSavingsMsg'
Learn More

Kif15-IN-1, MedChemExpress

Product Code. 16437006
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16437006

Brand: Medchem Express HY15948

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Kif15-IN-1 is an inhibitor of the mitotic Kinesin family member 15 (Kif15), and is used for the research of cellular proliferative diseases.

Kif15-IN-1 is an inhibitor of the mitotic Kinesin family member 15 (Kif15), and is used for the research of cellular proliferative diseases.

Specifications

Chemical Name or Material Kif15-IN-1
Physical Form Solid
CAS 672926-32-8
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C20H22N4O5S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 100 mg/mL (232.30 mM)
SMILES O=C(C1=CC=CC=C1N2)N([C@@H](C(C)C)C(NC3=NC(C)=C(C(OCC)=O)S3)=O)C2=O
Molecular Weight (g/mol) 430.48
Formula Weight 430.48
Percent Purity 99.23%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.