Learn More
Invitrogen™ KIF14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587769
Description
Immunogen sequence: LENGNNKAAS VQEEFMDAVC DGVGLGMKIL LDSGLEKAKE LQHELFRQCT KNEVTKEMKT NAMGLIRSLE NIFAESKIKS FRRQVQEENF EYQDFKRMVN RAPEFLKLKH CLEKAIEIII SALKGCHSDI NLLQTCVESI RNLASDFYSD FSVPSTSVGS YESRVTHIVH QELESLAKSL LFCFESEESP DLLKPWETYN QNTKEEHQQS KSSGIDGSKN KGVPKRVYEL HGSSPAVSSE ECTPSRIQWV; Positive Samples: HeLa, HL-60; Cellular Location: Cytoplasm, Midbody, Nucleus, cytoskeleton, spindle.
This gene encodes a member of the kinesin-3 superfamily of microtubule motor proteins. These proteins are involved in numerous processes including vesicle transport, chromosome segregation, mitotic spindle formation, and cytokinesis. In human HeLa-S3 and 293T cells, this protein is localized to the cytoplasm during interphase, to the spindle poles and spindle microtubules during mitosis, and to the midbody during cytokinesis. An internal motor domain displays microtubule-dependent ATPase activity, consistent with its function as a microtubule motor protein. Knockdown of this gene results in failed cytokinesis with endoreplication, which results in multinucleated cells. This gene has been identified as a likely oncogene in breast, lung and ovarian cancers, as well as retinoblastomas and gliomas. Alternative splicing results in multiple transcript variants.
Specifications
| KIF14 | |
| Polyclonal | |
| Unconjugated | |
| KIF14 | |
| D1Ertd367e; E130203M01; KIAA0042; KIF14; kinesin family member 14; kinesin-like protein KIF14; MKS12; N-3 kinesin; truncated kinesin family member 14 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 360849, 381293, 9928 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2.46 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| L0N7N1, Q15058 | |
| KIF14 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1399-1648 of human KIF14 (NP_0556901). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.