missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF14 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-55378
This item is not returnable.
View return policy
Description
KIF14 Polyclonal specifically detects KIF14 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| KIF14 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| KIAA0042MGC142302, kinesin family member 14, kinesin-like protein KIF14 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| KIF14 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MKLSMMIQEANAISSKLKTYYVFGRHDISDKSSSDTSIRVRNLKLGISTFWSLEKFESKLAAMKELYESNGSNRGEDA | |
| 100 μL | |
| Cell Biology, Cytoskeleton Markers, DNA Repair | |
| 9928 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction