missing translation for 'onlineSavingsMsg'
Learn More

KIF14 Antibody, Biotin, Novus Biologicals™

Product Code. 18419648 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419648 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419648 Supplier Novus Biologicals Supplier No. NB100254B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KIF14 Polyclonal antibody specifically detects KIF14 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF14
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot
Formulation PBS
Gene Alias KIAA0042MGC142302, kinesin family member 14, kinesin-like protein KIF14
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 900 and 950 of human Kinesin Family Member 14 using the numbering given in entry NP_055690.1 (GeneID 9928).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 9928
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.