missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF13B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | KIF13B |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18653557
|
Novus Biologicals
NBP2-62619-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18607158
|
Novus Biologicals
NBP2-62619 |
100 μg |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KIF13B Polyclonal antibody specifically detects KIF13B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| KIF13B | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| GAKIN, GAKINkinesin 13B, guanylate kinase associated kinesin, KIAA0639kinesin-like protein KIF13B, kinesin family member 13B, Kinesin-like protein GAKIN | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LPSGKNDGSIGGKQYFRCNPGYGLLVRPSRVRRATGPVRRRSTGLRLGAPEARRSATLSGSATNLASLTAALAKADRSHKNPENRKSWAS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| 23303 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title