missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KIF13A Polyclonal antibody specifically detects KIF13A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | KIF13A |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Formulation | PBS (pH 7.2) and 40% Glycerol |
| Gene Alias | bA500C11.2, FLJ27232, homolog of mouse KIF13A mannose-6-phosphate receptor transporter, kinesin family member 13A, kinesin-like protein KIF13A, Kinesin-like protein RBKIN, RBKIN |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: TGLPLNLSNFVFCQYTFWDQCESTVAAPVVDPEVPSPQSKDAQYTVTFSHCKDYVVNVTEEFLEFISDGALAIEVWGHRCAGNGSSIWEVDSLHAK |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?