missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF13A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35615-100ul
This item is not returnable.
View return policy
Description
KIF13A Polyclonal antibody specifically detects KIF13A in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| KIF13A | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| bA500C11.2, FLJ27232, homolog of mouse KIF13A mannose-6-phosphate receptor transporter, kinesin family member 13A, kinesin-like protein KIF13A, Kinesin-like protein RBKIN, RBKIN | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1300-1600 of human KIF13A (NP_001099036.1).,, Sequence:, SCGVTYEIVSNIPKATEEIEDRETLALLAARSENEGTSDGETYIEKYTRGVLQVENILSLERLRQAVTVKEALSTKARHIRRSLSTPNVHNVSSSRPDLSGFDEDDKGWPENQLDMSDYSSSYQDVACYGTLPRDSPRRNKEGCTSETPHALTVSPFKAFSPQPPKFFKPLMPVKEEHKKRIALEARPLLSQEDSEEEENELEAINRKLISSQPYVPVEFADFSVYNASLENREWFSSKVDLSNSRVLEKEVSRSPTTSSITSGYFSHSASNATLSDMVVPSSDSSDQLAIQTKDADSTEH | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 63971 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction