missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KIDINS220 Monoclonal Antibody (P220 1F8/3)

Product Code. 11590982
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11590982 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11590982 Supplier Invitrogen™ Supplier No. MA190667

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-90667 detects Kidins220 from rat samples.

This gene encodes a transmembrane protein that is preferentially expressed in the nervous system where it controls neuronal cell survival, differentiation into exons and dendrites, and synaptic plasticity. The encoded protein interacts with membrane receptors, cytosolic signaling components, and cytoskeletal proteins, serving as a scaffold that mediates crosstalk between the neurotrophin pathway and several other intracellular signaling pathways. Aberrant expression of this gene is associated with the onset of various neuropsychiatric disorders and neurodegenerative diseases, including Alzheimer's disease. Naturally occurring mutations in this gene are associated with a syndrome characterized by spastic paraplegia, intellectual disability, nystagmus and obesity. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIDINS220
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone P220 1F8/3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Kidins220
Gene Accession No. Q9EQG6
Gene Alias 3110039L19Rik; AI194387; AI316525; ankyrin repeat-rich membrane spanning; ankyrin repeat-rich membrane spanning protein; ankyrin repeat-rich membrane-spanning protein; ARMS; C330002I19Rik; KIAA1250; KIDINS220; kinase D-interacting substance 220; kinase D-interacting substance of 220 kDa; kinase D-interacting substrate 220; kinase D-interacting substrate 220kDa; kinase D-interacting substrate of 220 kDa; kinase D-interacting substrate, 220kDa; MGC163482; mKIAA1250; truncated KIDINS220
Gene Symbols Kidins220
Host Species Mouse
Immunogen Recombinant Kidins220/ARMS (last 347 amino acids at the carboxy terminus) protein.
Purification Method IgG fraction
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116478
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.