missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIDINS220 Monoclonal Antibody (3-B6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA532869
This item is not returnable.
View return policy
Description
KIDINS220 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a transmembrane protein that is preferentially expressed in the nervous system where it controls neuronal cell survival, differentiation into exons and dendrites, and synaptic plasticity. The encoded protein interacts with membrane receptors, cytosolic signaling components, and cytoskeletal proteins, serving as a scaffold that mediates crosstalk between the neurotrophin pathway and several other intracellular signaling pathways. Aberrant expression of this gene is associated with the onset of various neuropsychiatric disorders and neurodegenerative diseases, including Alzheimer's disease. Naturally occurring mutations in this gene are associated with a syndrome characterized by spastic paraplegia, intellectual disability, nystagmus and obesity. Alternative splicing results in multiple transcript variants.
Specifications
| KIDINS220 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9ULH0 | |
| Kidins220 | |
| Recombinant protein with Human kinase D-interacting substrate 220 aa 1601-1771. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 3-B6 | |
| Unconjugated | |
| Kidins220 | |
| 3110039L19Rik; AI194387; AI316525; ankyrin repeat-rich membrane spanning; ankyrin repeat-rich membrane spanning protein; ankyrin repeat-rich membrane-spanning protein; ARMS; C330002I19Rik; KIAA1250; KIDINS220; kinase D-interacting substance 220; kinase D-interacting substance of 220 kDa; kinase D-interacting substrate 220; kinase D-interacting substrate 220kDa; kinase D-interacting substrate of 220 kDa; kinase D-interacting substrate, 220kDa; MGC163482; mKIAA1250; truncated KIDINS220 | |
| Mouse | |
| Protein A | |
| RUO | |
| 57498, 77480 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction