missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KID Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583533
This item is not returnable.
View return policy
Description
Immunogen sequence: SLGGSAHSIL IANIAPERRF YLDTVSALNF AARSKEVINR PFTNESLQPH ALGPVKLSQK ELLGPPEAKR ARG Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 89%.
The protein encoded by this gene is a member of kinesin-like protein family. This family of proteins are microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. The C-terminal half of this protein has been shown to bind DNA. Studies with the Xenopus homolog suggests its essential role in metaphase chromosome alignment and maintenance.
Specifications
| KID | |
| Polyclonal | |
| Unconjugated | |
| KIF22 | |
| A-328A3.2; AU021460; C81217; Kid; Kif22; Kif22a; kinesin family member 22; kinesin family member 22A; kinesin-like 4; Kinesin-like DNA-binding protein; kinesin-like DNA-binding protein pseudogene; kinesin-like protein 4; Kinesin-like protein KIF22; KNSL4; OBP; OBP-1; OBP-2; origin of plasmid DNA replication-binding protein; oriP binding protein; SEMDJL2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3835 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14807 | |
| KIF22 | |
| Recombinant protein corresponding to Human KIF22. Recombinant protein control fragment (Product #RP-98673). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction