missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KID Monoclonal Antibody (5F3)

Product Code. 12817932
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12817932 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12817932 Supplier Invitrogen™ Supplier No. MA515910

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA5-15910 targets KID in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15910 immunogen is purified recombinant fragment of human KID expressed in E. Coli. . MA5-15910 detects KID which has a predicted molecular weight of approximately 73kDa.

The protein encoded by this gene is a member of kinesin-like protein family. This family of proteins are microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. The C-terminal half of this protein has been shown to bind DNA. Studies with the Xenopus homolog suggests its essential role in metaphase chromosome alignment and maintenance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KID
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5F3
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene KIF22
Gene Accession No. Q14807
Gene Alias A-328A3.2; AU021460; C81217; Kid; Kif22; Kif22a; kinesin family member 22; kinesin family member 22A; kinesin-like 4; Kinesin-like DNA-binding protein; kinesin-like DNA-binding protein pseudogene; kinesin-like protein 4; Kinesin-like protein KIF22; KNSL4; OBP; OBP-1; OBP-2; origin of plasmid DNA replication-binding protein; oriP binding protein; SEMDJL2
Gene Symbols KIF22
Host Species Mouse
Immunogen Purified recombinant fragment of human KID expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3835
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.