missing translation for 'onlineSavingsMsg'
Learn More

KIBRA Antibody (OTI1H8), DyLight 594, Novus Biologicals™

Product Code. 18407115 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407115 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407115 Supplier Novus Biologicals Supplier No. NBP271732DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KIBRA Monoclonal antibody specifically detects KIBRA in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIBRA
Applications Western Blot
Classification Monoclonal
Clone OTI1H8
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias FLJ10865, FLJ23369, HBeAg-binding protein 3, KIAA0869HBEBP3, KIBRAHBEBP36, Kidney and brain protein, protein KIBRA, protein WWC1, WW and C2 domain containing 1, WW domain-containing protein 1, WW, C2 and coiled-coil domain containing 1
Host Species Mouse
Immunogen Full length recombinant protein of human KIBRA (NP_056053) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 23286
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.