missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIBRA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35721-100ul
This item is not returnable.
View return policy
Description
KIBRA Polyclonal antibody specifically detects KIBRA in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| KIBRA | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| FLJ10865, FLJ23369, HBeAg-binding protein 3, KIAA0869HBEBP3, KIBRAHBEBP36, Kidney and brain protein, protein KIBRA, protein WWC1, WW and C2 domain containing 1, WW domain-containing protein 1, WW, C2 and coiled-coil domain containing 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 290-426 of human KIBRA (NP_056053.1).,, Sequence:, FGINSNNQLAEKVRLRLRYEEAKRRIANLKIQLAKLDSEAWPGVLDSERDRLILINEKEELLKEMRFISPRKWTQGEVEQLEMARKRLEKDLQAARDTQSKALTERLKLNSKRNQLVRELEEATRQVATLHSQLKSL | |
| 100 μL | |
| Neuroscience | |
| 23286 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction