missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KIAA1755 Recombinant Mouse Monoclonal Antibody (K52071_6D12)
GREENER_CHOICE

Product Code. 30829320
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829320 100 μL
30829604 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829320 Supplier Invitrogen™ Supplier No. MA564441

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: SREAGTSSGC TSGALEEIAG TKETPLFQKI LPLSEANEGP SLGNRACTKP ESSEERPYNL GFRRKVNLKA PTHNSERPPQ GSYMNVLEDA LDCASGLRAG VSQEPAASKM QGPLGNPENM VQLRPGPRQA SSPRLSPASP AAAASETKIE VKTKERNGRL Highest antigen sequence identity to the following orthologs: Mouse - 48%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1755
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone K52071_6D12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide
Gene KIAA1755
Gene Accession No. Q5JYT7
Gene Alias KIAA1755; RP5-1054A22.3; uncharacterized protein KIAA1755
Gene Symbols KIAA1755
Host Species Mouse
Immunogen Recombinant human KIAA1755 (aa 291-450).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85449
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.