Learn More
Invitrogen™ KIAA1751 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA571023
Product Code. 15839433
  • £441.00 / 100µL
Description
This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 100%; Human: 100%; Rat: 90%.
As part of the central apparatus of the cilium axoneme may play a role in cilium movement. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KIAA1751
Polyclonal
Unconjugated
CFAP74
C1orf222; CFAP74; cilia and flagella associated protein 74; cilia- and flagella-associated protein 74; FLJ45476; hCG1647286-like; KIAA1751; LOW QUALITY PROTEIN: cilia- and flagella-associated protein 74
Rabbit
Affinity Chromatography
RUO
85452
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9C0B2
CFAP74
synthetic peptide directed towards the C-terminal region of human C1orf222.