missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIAA1680 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559770
This item is not returnable.
View return policy
Description
Immunogen sequence: KEHSTNKNVF INCLSSGKSE GDDSGFTEDQ TRRSVKQSTR KLLPKSFSSH YKFSKPVLQS QSISLVQQSE FSLEVTQYQE REPVLVRASP SCSVDVTERA Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 81%.
Specifications
| KIAA1680 | |
| Polyclonal | |
| Unconjugated | |
| CCSER1 | |
| CCSER1; coiled-coil serine rich protein 1; coiled-coil serine-rich protein 1; FAM190A; family with sequence similarity 190, member A; KIAA1680; protein FAM190A; serine-rich coiled-coil domain-containing protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 401145 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9C0I3 | |
| CCSER1 | |
| Recombinant protein corresponding to Human KIAA1680. Recombinant protein control fragment (Product #RP-100265). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction