missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIAA1486 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569177
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%; Zebrafish: 100%.
Activates PI3K and concomitantly recruits the WAVE1 complex to the close vicinity of PI3K and regulates neuronal morphogenesis.
Specifications
| KIAA1486 | |
| Polyclonal | |
| Unconjugated | |
| NYAP2 | |
| 9430031J16Rik; Jr6; Kiaa1486; neuronal tyrosine phosphorylated adaptor for phosphoinositide 3-kinase adaptor 2; neuronal tyrosine-phophorylated phosphoinositide 3-kinase adaptor 2; neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adapter 2; neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adaptor 2; NEWGENE_1305560; NYAP2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57624 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9P242 | |
| NYAP2 | |
| synthetic peptide directed towards the middle region of human KIAA1486. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction