missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA1429 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25712-1-AP-20UL
This item is not returnable.
View return policy
Description
EIF2AK4 (GCN2) is a metabolic-stress sensing protein kinase that phosphorylates the alpha subunit of eukaryotic translation initiation factor-2 in response to low amino acid availability. It plays a role as an activator of the integrated stress response required for adapatation to amino acid starvation. Mutations in the gene lead to pulmonary vennocclusive disease 2.Specifications
| KIAA1429 | |
| Polyclonal | |
| Unconjugated | |
| KIAA1429 | |
| fSAP121, KIAA1429, MSTP054, Protein virilizer homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25962 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q69YN4 | |
| KIAA1429 | |
| KIAA1429 Fusion Protein Ag22449 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction