missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIAA1429 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107046
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
KIAA1429 is required for N6-methyladenosine (m6A) methylation of mRNAs, a modification that plays a role in the efficiency of mRNA splicing and processing. It is also Involved in mRNA splicing regulation, probably via its function in m6A methylation. Diseases associated with KIAA1429 include Retinitis Pigmentosa 42.
Specifications
| KIAA1429 | |
| Polyclonal | |
| Unconjugated | |
| VIRMA | |
| 1110037F02Rik; 4930422M05Rik; DKFZP434I116; fSAP121; functional spliceosome-associated protein 121; KIAA1429; mKIAA1429; MSTP054; protein virilizer homolog; RGD1559904; RIKEN cDNA 1110037F02 gene; similar to mKIAA1429 protein; Virma | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 25962 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q69YN4 | |
| VIRMA | |
| Synthetic peptide directed towards the C-terminal region of Human KIAA1429. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction