missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA1429 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68235-1-IG-20UL
This item is not returnable.
View return policy
Description
The KIAA1429 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human KIAA1429. This antibody recognizes human, mouse antigen. The KIAA1429 antibody has been validated for the following applications: WB, FC (Intra), ELISA analysis.Specifications
| KIAA1429 | |
| Monoclonal | |
| 500 μg/mL | |
| DKFZp434I116; DKFZp781B2117; MGC138493; MGC141940; MSTP054; fSAP121 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| 200-210 kDa | |
| Liquid |
| Western Blot, Flow Cytometry (Intracellular Staining), ELISA | |
| 3F1A4 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 25962 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction