missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIAA1328 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559365
This item is not returnable.
View return policy
Description
Immunogen sequence: QEELHMKECP HLKPTPSQCC GHRLAADRVH DSHPTNMTPQ HPKTHPESCS YCRLSWASLV HGGGALQPIE TLKKQISEDR KQQLMLQKME LE Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 63%.
Competes with SMC1 for binding to SMC3. May affect the availability of SMC3 to engage in the formation of multimeric protein complexes.
Specifications
| KIAA1328 | |
| Polyclonal | |
| Unconjugated | |
| KIAA1328 | |
| 4921538P22; AI461742; AW554918; expressed sequence AW554918; hinderin; KIAA1328; mKIAA1328; Protein hinderin; uncharacterized protein KIAA1328; uncharacterized protein KIAA1328; protein hinderin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57536 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86T90 | |
| KIAA1328 | |
| Recombinant protein corresponding to Human KIAA1328. Recombinant protein control fragment (Product #RP-98254). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction