missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KIAA1324 Polyclonal Antibody

Product Code. 15724191 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15724191 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15724191 Supplier Invitrogen™ Supplier No. PA556277

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SLCYNDCTFS RNTPTRTFNY NFSALANTVT LAGGPSFTSK GLKYFHHFTL SLCGNQGRKM SVCTDNVTDL RIPEGESGFS KSITAYVCQA VIIPPEVTGY KAGVSSQPVS LADRLIGVTT DMTLDGITSP AELFHLESLG IPDVIF Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 86%.

Expression of this gene is induced by estrogen and the encoded protein has been characterized as a transmembrane protein. The encoded protein has been found in to correlate with survival in certain carcinomas (PMID: 21102415) and may be important for cellular response to stress (PMID: 21072319). Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1324
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIAA1324
Gene Accession No. Q6UXG2
Gene Alias EIG121; Estrogen-induced gene 121 protein; KIAA1324; maba1; UNQ2426/PRO4985; UPF0577 protein KIAA1324
Gene Symbols KIAA1324
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIAA1324. Recombinant protein control fragment (Product #RP-88751).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57535
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.