Learn More
KIAA1310 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557116
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PVLFVIGQNS LQCHPEAMED FREKIRAENS LVVVGGADDN LRISKAKKKS EGLTQSMVDR CIQDEIVDFL TGVLTRAEGH MGSEPRDQDA EKKKKPR Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
As part of the NSL complex it is involved in acetylation of nucleosomal histone H4 on several lysine residues and therefore may be involved in the regulation of transcription.Specifications
KIAA1310 | |
Polyclonal | |
Unconjugated | |
KANSL3 | |
KANSL3; KAT8 regulatory NSL complex subunit 3; KIAA1310; Non-specific lethal 3 homolog; NSL complex protein NSL3; NSL3; PRTD; Rcd1; serum inhibited-related protein; SI1; Testis development protein PRTD | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55683 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P2N6 | |
KANSL3 | |
Recombinant protein corresponding to Human KIAA1310 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only