missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA1279, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00026128-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MANVPWAEVCEKFQAALALSRVELHKNPEKEPYKSKYSARALLEEVKALLGPAPEDEDERPEAEDGPGAGDHALGLPAEVVEPEGPVAQRAVRLAVIEFHLGVNHIDTEELSAGEEHLVKCLRLLRRYRLSHDCISLCIQAQNNLGILWSEREEIETAQAYLESSEALYNQYMKEVGSPPLDPTERFLPEEEKLTEQERSKRFEKVYTHNLYYLAQVYQHLEMFEKAAHYCHSTLKRQLEHNAYHPIEWAINAATLSQFYINKLCFMEARHCLSAANVIFGQTGKISATEDTPEAEGEVPELYHQRKGEIARCWIKYCLTLMQNAQLSMQDNIGELDLDKQSELRALRKKELDEEESIRKKAVQFGTGELCDAISAVEEKVSYLRPLDFEEARELFLLGQHYVFEAKEFFQIDGYVTDHIEVVQDHSALFKVLAFFETDMERRCKMHKRRIAMLEPLTVDLNPQYYLLVNRQIQFEIAHAYYDMMDLKVAIADRLRDPDSHIVKKINNLNKSALKYYQLFLDSLRDPNKVFPEHIGEDVLRPAMLAKFRVARLYGKIITADPKKELENLATSLEHYKFIVDYCEKHPEAAQEIEVELELSKEMVSLLPTKMERFRTKMALTSpecifications
| KIAA1279 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_015634 | |
| KIAA1279 | |
| KIAA1279 (NP_056449, 1 a.a. ∼ 621 a.a) full-length human protein. | |
| RUO | |
| 26128 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| KIAA1279 | |
| DKFZp586B0923/KBP/TTC20 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction