missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIAA1199 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145216
This item is not returnable.
View return policy
Description
Immunogen sequence: RVFKLFQTEH GEYFNVSLSS EWVQDVEWTE WFDHDKVSQT KGGEKISDLW KAHPGKICNR PIDIQATTMD GVNLSTEVVY KKGQDYRFAC YDRGRACRSY RVRFLCGKPV RPKLTVTIDT NVNSTILNLE DNVQSWKPGD TLVIA Antigen sequence identity: Mouse 89%; Rat 88%.
The function of this protein remains unknown.
Specifications
| KIAA1199 | |
| Polyclonal | |
| Unconjugated | |
| Cemip | |
| 12H19.01.T7; 6330404C01Rik; 9930013L23Rik; AY007814; CCSP1; cell migration inducing hyaluronan binding protein; cell migration inducing hyaluronidase 1; cell migration inducing protein, hyaluronan binding; cell migration-inducing and hyaluronan-binding protein; CEMIP; colon cancer secreted protein 1; gb:bi851057; hyaluronan-binding protein involved in hyaluronan depolymerization; HYBID; Kiaa1199; Protein 12H19.01.T7; protein KIAA1199; RGD1305254; TMEM2L | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 57214 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WUJ3 | |
| Cemip | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 298-442. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction