missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA1107 Polyclonal Antibody

KIAA1107 Polyclonal Antibody
Brand: Thermo Scientific PA555017
This item is not returnable.
View return policy
Description
KIAA1107 Polyclonal Antibody for IHC
Specifications
| KIAA1107 | |
| Polyclonal | |
| Unconjugated | |
| BTBD8 | |
| BTB (POZ) domain containing 8; BTB domain containing 8; BTB/POZ domain-containing protein 8; BTBD8; double BTB/POZ domain containing protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 284697 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5XKL5 | |
| BTBD8 | |
| Recombinant protein corresponding to Human KIAA1107. Recombinant protein control fragment (Product #RP-94010). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction