missing translation for 'onlineSavingsMsg'
Learn More

KIAA0513 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen KIAA0513
Concentration 0.25 mg/mL
Applications Immunofluorescence, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16891065
View Documents
Proteintech
21478-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16881065
View Documents
Proteintech
21478-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The function of this protein remains unknown.
Specifications

Specifications

KIAA0513
Immunofluorescence, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human, Mouse, Rat
O60268, Q8R0A7
234797, 307907, 9764
KIAA0513 Fusion Protein Ag15624
Primary
-20°C
KIAA0513
0.25 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
KIAA0513
KIAA0513, RGD1304884
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.