missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIAA0391 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576469
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
KIAA0391, also known as MRPP3 (mitochondrial ribonuclease P protein 3), is a 583 amino acid protein that is located in the mitochondria. KIAA0391 functions in tRNA maturation and is a component of mitochondrial ribonuclease P which cleaves tRNA at the 5'-end. Mitochondrial ribonuclease P is an enzyme composed of RG9MTD1, MRPP22 and KIAA0391 subunits. Due to alternative splicing events, KIAA0391 exists as four isoforms. The gene encoding KIAA0391 maps to human chromosome 14, which contains about 700 genes and makes up about 3. 5% of human cellular DNA.
Specifications
| KIAA0391 | |
| Polyclonal | |
| Unconjugated | |
| PRORP | |
| 1110008L16Rik; KIAA0391; Mitochondrial ribonuclease P catalytic subunit; Mitochondrial ribonuclease P protein 3; mitochondrial RNase P protein 3; mitochondrial RNase P subunit 3; mKIAA0391; MRPP3; Prorp; Protein only RNase P catalytic subunit; proteinaceous RNase P; RGD1305089; RIKEN cDNA 1110008L16 gene; similar to 1110008L16Rik protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 299050, 9692 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| B5DF07, O15091 | |
| PRORP | |
| Recombinant fusion protein of human KIAA0391. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction