missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA0319L Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21016-1-AP-20UL
This item is not returnable.
View return policy
Description
Possible role in axon guidance through interaction with RTN4R. (Microbial infection) Acts as a receptor for adeno-associated virus and is involved in adeno-associated virus infection through endocytosis system.Specifications
| KIAA0319L | |
| Polyclonal | |
| Unconjugated | |
| KIAA0319L | |
| KIAA0319 like, KIAA0319L, KIAA1837, PKD1 like, PP791, RP4 765A10.3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100317, 79932 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8IZA0, Q8K135 | |
| KIAA0319L | |
| KIAA0319L Fusion Protein Ag15263 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction